Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102669 Supplier Invitrogen™ Supplier No. PA5102669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HOXC8.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HOXC8
Gene Accession No. P09025, P18866, P31273
Gene Alias D130011F21Rik; homeo box 3A; homeo box C8; homeobox C8; Homeobox gene C8; homeobox protein Hox-3.1; Homeobox protein Hox-3A; homeobox protein Hox-C8; Homeobox protein M31; homeobox protein R4; HOX3; Hox-3.1; Hox-3.1, mouse, homolog of; HOX3A; Hox3r4; HOXC8; Hoxc-8
Gene Symbols HOXC8
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXC8(Accession P31273), corresponding to amino acid residues S78-H128.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15426, 24460, 3224
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.