Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567225
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567225 Supplier Invitrogen™ Supplier No. PA567225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSQSSVVYHPY GPQPHLGADT RYMRTWLEPL SGAVSFPSFP AGGRHYALKP DAY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

HOXC9 belongs to the homeobox family. The homeobox family is a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC9
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXC9
Gene Accession No. P31274
Gene Alias homeo box C9; homeobox C9; homeobox protein Hox-3.2; Homeobox protein Hox-3B; Homeobox protein Hox-C9; HOX3; Hox-3.2; HOX3B; Hoxc8; Hoxc9; HOXC-9
Gene Symbols HOXC9
Host Species Rabbit
Immunogen Recombinant Human HOXC9. Recombinant protein control fragment (Product #RP-107863).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3225
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.