Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541125 Supplier Invitrogen™ Supplier No. PA541125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DRAPQTLPSP EADALAGSKH KEEKADLDPS NPVANWIHAR STRKKRCPYT Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 79%; Rabbit: 93%; Rat: 100%; Sheep: 100%.

HOXC9 belongs to the homeobox family. The homeobox family is a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOXC9
Gene Accession No. P31274
Gene Alias homeo box C9; homeobox C9; homeobox protein Hox-3.2; Homeobox protein Hox-3B; Homeobox protein Hox-C9; HOX3; Hox-3.2; HOX3B; Hoxc8; Hoxc9; HOXC-9
Gene Symbols HOXC9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HOXC9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3225
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.