Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566047 Supplier Invitrogen™ Supplier No. PA566047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REREGLLATAI PVAPLQLPLS GTTPTKFIKN PGSPSQSQEP S Highest antigen sequence identity to the following orthologs - mouse 67%, rat 71%.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA1 is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXD1
Gene Accession No. Q9GZZ0
Gene Alias BSAS; homeo box 4G; homeo box A1; homeo box D1; homeobox 1F; homeobox A1; homeobox D1; homeobox protein Hox-4.9; Homeobox protein Hox-D1; homeobox protein Hox-GG; HOX1; HOX1F; HOX2I; HOX4; Hox-4.7; Hox-4.9; HOX4G; HOXA1; Hoxd1; Hoxd-1; MGC45232
Gene Symbols HOXD1
Host Species Rabbit
Immunogen Recombinant Human HOXD1. Recombinant protein control fragment (Product #RP-105058).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.