Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113456
Description
HOXD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA1 is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region.
Specifications
| HOXD1 | |
| Polyclonal | |
| Unconjugated | |
| HOXD1 | |
| BSAS; homeo box 4G; homeo box A1; homeo box D1; homeobox 1F; homeobox A1; homeobox D1; homeobox protein Hox-4.9; Homeobox protein Hox-D1; homeobox protein Hox-GG; HOX1; HOX1F; HOX2I; HOX4; Hox-4.7; Hox-4.9; HOX4G; HOXA1; Hoxd1; Hoxd-1; MGC45232 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 15429, 288151, 3231 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q01822, Q9GZZ0 | |
| HOXD1 | |
| Recombinant Human Homeobox protein Hox-D1 protein (59-131AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction