Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD10 Antibody (OTI1D11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270920F
Description
HOXD10 Monoclonal specifically detects HOXD10 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
HOXD10 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HOXD10 | |
Recombinant protein fragment corresponding to amino acids 80-340 of human HOXD10 (NP_002139) produced in E.coli. | |
0.1 mL | |
3236.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI1D11 | |
Immunohistochemistry | |
homeo box 4D, homeo box D10, homeobox D10, Homeobox protein Hox-4D, Homeobox protein Hox-4E, homeobox protein Hox-D10, Hox-4.4, HOX4DHOX4, HOX4E | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction