Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXD10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546861
Description
Peptide sequence: NPEVPVPGYF RLSQTYATGK TQEYNNSPEG SSTVMLQLNP RGAAKPQLSA Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 92%.
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.
Specifications
HOXD10 | |
Polyclonal | |
Unconjugated | |
HOXD10 | |
AI385591; AI874987; d10; homeo box 4D; homeo box D10; homeo box D10a; homeobox D10; homeobox D10a; homeobox gene D-10; homeobox protein Hox-4.5; Homeobox protein Hox-4D; homeobox protein Hox-4E; Homeobox protein Hox-5.3; homeobox protein Hox-C10; Homeobox protein Hox-D10; homeobox protein Hox-D10a; homeobox protein Hox-D11; homeobox protein Hox-D10; HOX4; Hox-4.4; Hox-4.5; Hox-45; HOX4D; HOX4E; Hox-5.3; hoxc10; hox-C10; Hoxc10 protein; Hoxd10; HOX-D10; hoxd-10; hoxd10a; hoxd10a protein; z-82 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3236 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P28358 | |
HOXD10 | |
Synthetic peptide directed towards the middle region of human HOXD10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction