Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567572 Supplier Invitrogen™ Supplier No. PA567572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene HOXD10
Gene Accession No. P28358, P28359
Gene Alias AI385591; AI874987; d10; homeo box 4D; homeo box D10; homeo box D10a; homeobox D10; homeobox D10a; homeobox gene D-10; homeobox protein Hox-4.5; Homeobox protein Hox-4D; homeobox protein Hox-4E; Homeobox protein Hox-5.3; homeobox protein Hox-C10; Homeobox protein Hox-D10; homeobox protein Hox-D10a; homeobox protein Hox-D11; homeobox protein Hox-D10; HOX4; Hox-4.4; Hox-4.5; Hox-45; HOX4D; HOX4E; Hox-5.3; hoxc10; hox-C10; Hoxc10 protein; Hoxd10; HOX-D10; hoxd-10; hoxd10a; hoxd10a protein; z-82
Gene Symbols HOXD10
Host Species Rabbit
Immunogen Synthesized peptide derived from human HOXD10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15430, 3236
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.