Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | HOXD10 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HOXD10 Polyclonal specifically detects HOXD10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
HOXD10 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3236 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
homeo box 4D, homeo box D10, homeobox D10, Homeobox protein Hox-4D, Homeobox protein Hox-4E, homeobox protein Hox-D10, Hox-4.4, HOX4DHOX4, HOX4E | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human HOXD10 (NP_002139.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title