Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116229
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116229 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116229 Supplier Invitrogen™ Supplier No. PA5116229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXD11 Polyclonal Antibody for Western Blot

HOX11, also called homeobox11 or Tcl-3 is located at the 10q24 T-Cell translocation breakpoint region. It encodes a homeobox-domain containing protein, HOX11, containing a glycine and proline-rich amino terminus. HOX11 is required for maintenance of the developing spleen and cell survival. The HOX11protein binds to a specific DNA sequence and localizes to the cell nucleus. It transactivates transcription of a reporter gene linked to a cis-regulatory element, acting as a positive transcription activator. The HOX11 gene is highly expressed in T-ALLs as a result of a t (7:10) (q35;q24) or t(10:14)(q24;qll) translocation. The homeobox gene is deregulated upon translocation into the T-cell receptor locus and activates a specific subset of tumor-associated target genes. HOX 11 gene translocation has been attributed to T-cell leukemia and lymphoid neoplasias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HOXD11
Gene Accession No. P31277
Gene Alias E230017H14Rik; homeo box 4F; homeo box D11; homeobox D11; homeobox protein Hox-4.6; Homeobox protein Hox-4F; homeobox protein Hox-5.5; Homeobox protein Hox-D11; HOX11; HOX4; Hox-4.6; Hox-4.6, mouse, homolog of; HOX4F; Hox-5.4; Hox-5.5; Hoxd11; Hoxd-11; LOC102553842; uncharacterized LOC102553842
Gene Symbols HOXD11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 187-216 amino acids from the Central region of human HOXD11.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.