Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD12 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP226178
Description
HOXD12 Polyclonal specifically detects HOXD12 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
HOXD12 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
HOXD12 | |
Peptide with sequence C-PTAKDGPEEQAK corresponding to internal region according to NP_067016.3. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 2 - 3 ug/ml, Peptide ELISA Detection limit 1:1000 | |
homeobox D12, Homeobox protein Hox-4H, mouse, homolog of | |
Goat | |
Affinity Purified | |
RUO | |
3238 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction