Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599289
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599289 Supplier Invitrogen™ Supplier No. PA599289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HOXD12.

The Hox proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. HoxD12 (homeobox D12), also known as HOX4H, is a 275 amino acid protein that localizes to the nucleus and contains one homeobox DNA-binding domain. One of several members of the homeobox superfamily, HoxD12 functions as a sequencespecific transcription factor that is important for the correct positioning of developing limb buds on the anterior-posterior axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HOXD12
Gene Accession No. P23812, P35452
Gene Alias homeo box D12; homeobox D12; homeobox protein Hox-4.7; Homeobox protein Hox-4H; homeobox protein Hox-5.6; Homeobox protein Hox-D12; Hox 4H; Hox-4.7; Hox-4.7, mouse, homolog of; HOX4H; Hox-5.6; Hoxd12; Hoxd-12
Gene Symbols HOXD12
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXD12(Accession P35452), corresponding to amino acid residues A68-S118.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15432, 3238
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.