Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568541 Supplier Invitrogen™ Supplier No. PA568541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.

HOXD13 (homeobox protein D13) belongs to a class of transcription factors called homeobox proteins. In vertebrates, homeobox are found in clusters called A, B, C, and D on four separate chromosomes. Expression of homeobox proteins is spatially and temporally regulated during embryonic development. The gene encoding HOXD13 is part of the D cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. Mutations in the HOXD13 gene cause synpolydactyly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOXD13
Gene Accession No. P70217
Gene Alias BDE; BDSD; homeo box 4I; homeo box D13; homeobox D13; homeobox protein Hox-4.8; Homeobox protein Hox-4I; homeobox protein Hox-D13; Hox-4.8; HOX4I; Hoxd13; SPD; SPD1; spdh
Gene Symbols HOXD13
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse HOXD13 (aa 174-223).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15433
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.