Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXD13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575937
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 96% (by SDS-PAGE).
HOXD13 (homeobox protein D13) belongs to a class of transcription factors called homeobox proteins. In vertebrates, homeobox are found in clusters called A, B, C, and D on four separate chromosomes. Expression of homeobox proteins is spatially and temporally regulated during embryonic development. The gene encoding HOXD13 is part of the D cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. Mutations in the HOXD13 gene cause synpolydactyly.
Specifications
HOXD13 | |
Polyclonal | |
Unconjugated | |
HOXD13 | |
BDE; BDSD; homeo box 4I; homeo box D13; homeobox D13; homeobox protein Hox-4.8; Homeobox protein Hox-4I; homeobox protein Hox-D13; Hox-4.8; HOX4I; Hoxd13; SPD; SPD1; spdh | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15433, 288154, 3239 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P35453, P70217 | |
HOXD13 | |
Synthetic peptide corresponding to amino acids 173-218 of Human HOXD13. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction