Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXD13 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142544
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
HOXD13 (homeobox protein D13) belongs to a class of transcription factors called homeobox proteins. In vertebrates, homeobox are found in clusters called A, B, C, and D on four separate chromosomes. Expression of homeobox proteins is spatially and temporally regulated during embryonic development. The gene encoding HOXD13 is part of the D cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. Mutations in the HOXD13 gene cause synpolydactyly.
Specifications
| HOXD13 | |
| Polyclonal | |
| Unconjugated | |
| HOXD13 | |
| BDE; BDSD; homeo box 4I; homeo box D13; homeobox D13; homeobox protein Hox-4.8; Homeobox protein Hox-4I; homeobox protein Hox-D13; Hox-4.8; HOX4I; Hoxd13; SPD; SPD1; spdh | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 288154, 3239 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P35453 | |
| HOXD13 | |
| Peptide with sequence C-HGVGLQQNALKSSPH. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction