Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567561
Description
Predicted to react with Mouse samples.
HOXD3 (homeobox D3) belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that remove the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. HOXD3 may play a role in the regulation of cell adhesion processes.
Specifications
HOXD3 | |
Polyclonal | |
Unconjugated | |
Hoxd3 | |
homeo box D3; homeo box D4; homeobox D3; Homeobox gene D3; homeobox protein Hox-4.1; Homeobox protein Hox-4A; homeobox protein Hox-D3; Homeobox protein MH-19; homeobox protein R6; HOX1D; HOX4; Hox-4.1; Hox-4.1, mouse, homolog of; HOX4A; Hox4r6; Hox-5.5; HOXD3; Hoxd-3; MGC10470 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15434, 3232 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P09027, P31249 | |
Hoxd3 | |
Synthesized peptide derived from human HOXD3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction