Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590343 Supplier Invitrogen™ Supplier No. PA590343
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASQSPERSPP LGGAAGHVAY SGQLPPVPGL AYDAPSPPAF AKSQPNMYGL AAYTAPLSSC LPQQKRYAAP EFEPHPMASN GGGFASANLQ GSPVYVGGNF VESMAPASGP VFNLGHLSHP SSASVDYSCA AQIPGNHHHG PCDPHPTYTD LSAHHSSQGR LPEAPKLTHL; Positive Samples: Mouse liver, Mouse spinal cord, Rat spinal cord, Rat liver; Cellular Location: Nucleus.

HOXD3 (homeobox D3) belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that remove the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. HOXD3 may play a role in the regulation of cell adhesion processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Hoxd3
Gene Accession No. P09027, P31249
Gene Alias homeo box D3; homeo box D4; homeobox D3; Homeobox gene D3; homeobox protein Hox-4.1; Homeobox protein Hox-4A; homeobox protein Hox-D3; Homeobox protein MH-19; homeobox protein R6; HOX1D; HOX4; Hox-4.1; Hox-4.1, mouse, homolog of; HOX4A; Hox4r6; Hox-5.5; HOXD3; Hoxd-3; MGC10470
Gene Symbols Hoxd3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 263-432 of human HOXD3 (NP_0088293).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15434, 3232
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.