Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24963125UL
Description
HOXD4 Polyclonal antibody specifically detects HOXD4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
HOXD4 | |
Polyclonal | |
Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
HHO.C13, homeo box D4, homeobox D4, Homeobox protein HHO.C13, Homeobox protein Hox-4B, Homeobox protein Hox-5.1, homeobox protein Hox-D4, HOX4, Hox-4.2, Hox-4.2, mouse, homolog of homeo box X, HOX4BHOX-5.1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: ARAYSQSDPKQPPSGTALKQPAVVYPWMKKV | |
25 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
3233 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction