Learn More
Invitrogen™ HOXD4 Monoclonal Antibody (1H7)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524409
Catalog No. PIMA524409
Only null left
Add to Cart
Description
Description
Peptide Sequence: MVMSSYMVNS KYVDPKFPPC EEYLQGGYLG EQGADYYGGG AQGADFQPPG LYPRPDFGEQ PF.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HOXD4
Monoclonal
Unconjugated
HOXD4
6030436D05Rik; HHO.C13; homeo box D4; homeobox D4; homeobox protein HHO.C13; Homeobox protein Hox-4.2; homeobox protein Hox-4B; Homeobox protein Hox-5.1; homeobox protein Hox-D4; HOX4; Hox-4.2; Hox-4.2, mouse, homolog of homeo box X; HOX4B; Hox-5.1; HOXD4; Hoxd-4; LOW QUALITY PROTEIN: homeobox protein Hox-D4
Mouse
Protein A
RUO
3233
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1H7
PBS with no preservative; pH 7.4
P09016
HOXD4
HOXD4 (NP_055436.2, 1 a.a. ~ 62 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.