Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565867 Supplier Invitrogen™ Supplier No. PA565867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCEEYLQGGYL GEQGADYYGG GAQGADFQPP GLYPRPDFGE QPFGGSG Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXD4
Gene Accession No. P09016
Gene Alias 6030436D05Rik; HHO.C13; homeo box D4; homeobox D4; homeobox protein HHO.C13; Homeobox protein Hox-4.2; homeobox protein Hox-4B; Homeobox protein Hox-5.1; homeobox protein Hox-D4; HOX4; Hox-4.2; Hox-4.2, mouse, homolog of homeo box X; HOX4B; Hox-5.1; HOXD4; Hoxd-4; LOW QUALITY PROTEIN: homeobox protein Hox-D4
Gene Symbols HOXD4
Host Species Rabbit
Immunogen Recombinant Human HOXD4. Recombinant protein control fragment (Product #RP-106538).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.