Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568997
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
HOXD4 | |
Polyclonal | |
Unconjugated | |
HOXD4 | |
6030436D05Rik; HHO.C13; homeo box D4; homeobox D4; homeobox protein HHO.C13; Homeobox protein Hox-4.2; homeobox protein Hox-4B; Homeobox protein Hox-5.1; homeobox protein Hox-D4; HOX4; Hox-4.2; Hox-4.2, mouse, homolog of homeo box X; HOX4B; Hox-5.1; HOXD4; Hoxd-4; LOW QUALITY PROTEIN: homeobox protein Hox-D4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
288153 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
HOXD4 | |
synthetic peptide directed towards the N-terminal region of rat HOXD4. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction