Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597837
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597837 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597837 Supplier Invitrogen™ Supplier No. PA597837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXD8 Polyclonal Antibody for Western Blot, ELISA

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. In addition to effects during embryogenesis, this particular gene may also play a role in adult urogenital tract function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HOXD8
Gene Accession No. P13378, P23463
Gene Alias 4921540P06Rik; AI047735; homeo box 4E; homeo box D8; homeobox D8; homeobox protein 5.4; homeobox protein Hox-4.3; Homeobox protein Hox-4E; homeobox protein Hox-5.4; Homeobox protein Hox-D8; homeobox protein Hox-D8-like; HOX4; Hox-4.3; Hox-4.5; HOX4E; HOX5.4; Hox-5.4; HOXD8; Hoxd-8; LOC100912936
Gene Symbols HOXD8
Host Species Rabbit
Immunogen Recombinant Human Homeobox protein Hox-D8 protein (59-108AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15437, 3234
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.