Learn More
Invitrogen™ HOXD9 Monoclonal Antibody (OTI1H3)

Description
HOXD9 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | HOXD9 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1H3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HOXD9 |
| Gene Accession No. | P28356 |
| Gene Alias | homeo box D9; homeobox D9; homeobox Hox-D9; homeobox protein Hox-4.4; Homeobox protein Hox-4C; homeobox protein Hox-5.2; Homeobox protein Hox-D9; HOX4; Hox-4.3; Hox-4.3, mouse, homolog of; Hox-4.4; HOX4C; Hox-5.2; HOXD9; Hoxd-9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.