Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD9 Monoclonal Antibody (OTI4H10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809664

Description
HOXD9 is the sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5′ end of this cluster have been associated with severe limb and genital abnormalities. The exact role of this gene has not been determined.Specifications
HOXD9 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P28356 | |
HOXD9 | |
Human recombit protein fragment corresponding to amino acids 109-352 of human HOXD9 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4H10 | |
Unconjugated | |
HOXD9 | |
HOX4, HOX4C, Hox-4.3, Hox-5.2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3235 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction