Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540887 Supplier Invitrogen™ Supplier No. PA540887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RGPSPGPSGP ANGRHYGIKP ETRAAPAPAT AASTTSSSST SLSSSSKRTE Sequence homology: Cow: 92%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

HOXD9 is the sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. The exact role of this gene has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOXD9
Gene Accession No. P28356
Gene Alias homeo box D9; homeobox D9; homeobox Hox-D9; homeobox protein Hox-4.4; Homeobox protein Hox-4C; homeobox protein Hox-5.2; Homeobox protein Hox-D9; HOX4; Hox-4.3; Hox-4.3, mouse, homolog of; Hox-4.4; HOX4C; Hox-5.2; HOXD9; Hoxd-9
Gene Symbols HOXD9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HOXD9.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3235
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.