Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HP Mouse anti-Human, Clone: OTI1F9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA600399

Description
Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which aslo acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chanins bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.Specifications
HP | |
Monoclonal | |
0.5 mg/mL | |
PBS with <5% trehalose and 0.05% sodium azide | |
P00738 | |
HP | |
Full length human recombit protein of human HP produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
OTI1F9 | |
Unconjugated | |
HP | |
BP, HP2ALPHA2, HPA1S | |
Mouse | |
Affinity Chromatography | |
RUO | |
3240 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction