Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HP1 alpha Antibody (2H4E9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | HP1 alpha |
---|---|
Clone | 2H4E9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Description
HP1 alpha Monoclonal specifically detects HP1 alpha in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
HP1 alpha | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing | |
PBS with 0.05% Sodium Azide | |
23468 | |
Purified recombinant fragment of human CBX5 (AA: 1-191) expressed in E. Coli. | |
Primary | |
22.2 kDa |
2H4E9 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha | |
CBX5 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title