Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1 Alpha, Beta, and Gamma Antibody, Novus Biologicals™
SDP

Catalog No. NBP259154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259154 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259154 Supplier Novus Biologicals Supplier No. NBP259154
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HP1 Alpha, Beta, and Gamma Polyclonal specifically detects HP1 Alpha, Beta, and Gamma in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunofluorescence.

Specifications

Antigen HP1 Alpha, Beta, and Gamma
Applications Western Blot, Immunocytochemistry, Immunofluorescence, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 4 ug/IP
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias Antigen p25, CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 1, chromobox protein homolog 3, chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, Heterochromatin protein 1 homolog beta, Heterochromatin protein 1 homolog gamma, heterochromatin protein 1-beta, heterochromatin protein HP1 gamma, Heterochromatin protein p25, heterochromatin protein p25 beta, heterochromatin-like protein 1, HP1, HP1 alpha, HP1 alpha homolog, HP1 beta, HP1 beta homolog, HP1 gamma, HP1A, HP1-ALPHA, HP1-GAMMA, HP1Hs alpha, HP1Hs-alpha, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, Modifier 2 protein, p25beta
Host Species Rabbit
Immunogen The exact immunogen is propietary information.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23468
Target Species Human, Mouse
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.