Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HP1 alpha/beta/gamma Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA540091

Catalog No. PA540091


Only null left
Add to Cart

Description

Description

HP1 alpha/beta/gamma Polyclonal Antibody for Western Blot, ICC/IF

At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane. Two transcript variants encoding the same protein but differing in the 5' UTR, have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HP1 alpha/beta/gamma
Polyclonal
Unconjugated
CBX3
2610029O15Rik; Antigen p25; C75991; CBX; Cbx1; Cbx3; Cbx5; Cbx-rs2; chromobox 1; chromobox 3; chromobox 5; chromobox homolog 1; chromobox homolog 1 (HP1 beta homolog Drosophila ); chromobox homolog 3; chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox homolog 5; chromobox homolog 5 (HP1 alpha homolog, Drosophila); chromobox protein homolog 1; Chromobox protein homolog 3; chromobox protein homolog 5; E430007M08Rik; epididymis luminal protein 25; HECH; HEL25; heterochromatin protein 1 alpha; heterochromatin protein 1 gamma; heterochromatin protein 1 homolog alpha; Heterochromatin protein 1 homolog beta; heterochromatin protein 1 homolog gamma; heterochromatin protein 1-alpha; heterochromatin protein 1-beta; heterochromatin protein HP1 gamma; heterochromatin protein p25; heterochromatin protein p25 beta; heterochromatin-like protein 1; HP1; HP1 alpha; HP1 alpha homolog; HP1 beta; HP1 beta homolog; HP1 gamma; HP1 gamma homolog; HP1A; Hp1alpha; HP1-ALPHA; HP1B; Hp1beta; HP1-BETA; HP1g; HP1-GAMMA; HP1Hs alpha; HP1Hsbeta; HP1Hs-beta; HP1Hs-gamma; M31; M32; mHP1(alpha); MOD1; modifier 1 protein; Modifier 2 protein; p25beta
Rabbit
Protein G
RUO
10951, 11335, 12412, 12417, 12419, 23468
-20°C or -80°C if preferred
Liquid
Western Blot, Immunocytochemistry
2 mg/mL
PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4
P23198, P45973, P83916, P83917, Q13185, Q61686
CBX1, CBX3, CBX5
Human human HP1 beta (Heterochromatin protein 1 homolog beta).
50 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.