Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HP1 alpha Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142181
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142181 Supplier Invitrogen™ Supplier No. PA5142181
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Heterochromatin protein-1 (HP1) is a methyl-lysine binding protein localized at heterochromatin sites, where it mediates gene silencing. It has been shown that mammalian methyltransferases that selectively methylate histone H3 on lysine-9 generate a binding site for HP1 proteins, a family of heterochromatic adaptor molecules implicated in both gene silencing and supranucleosomal chromatin structure. High-affinity in vitro recognition of a methylated histone H3 peptide by HP1 requires a functional chromodomain. Thus, the HP1 chromodomain is a specific interaction motif for the methyl epitope on lysine-9 of histone H3. In vivo, heterochromatin association of HP1 proteins is lost in Suv39h double-null primary mouse fibroblasts but is restored after reintroduction of a catalytically active SUV39H1 HMTase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1 alpha
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Cbx5
Gene Accession No. P45973
Gene Alias 2610029O15Rik; antigen p25; C75991; Cbx5; Cbx5 antibody; chromobox 5; chromobox homolog 5; chromobox homolog 5 (HP1 alpha homolog, Drosophila); chromobox protein homolog 5; chromobox-like protein 5; epididymis luminal protein 25; HEL25; heterochromatin protein 1 alpha; heterochromatin protein 1 homolog alpha; heterochromatin protein 1-alpha; HGNC:1555; HP1; HP1 alpha; HP-1 alpha antibody; HP1 alpha homolog; Hp1a; Hp1alpha; HP1-ALPHA; HP1Hs alpha; HP1Hs-alpha; mHP1(alpha)
Gene Symbols Cbx5
Host Species Goat
Immunogen Peptide with sequence C-NKRKSNFSNSADDIK (aa 88-102).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23468
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.