Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HP1 beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145003 Supplier Invitrogen™ Supplier No. PA5145003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERLTWHSYPS EDDDKKDDKN Antigen sequence identity: Mouse 100%; Rat 100%.

This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1 beta
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CBX1
Gene Accession No. P83916
Gene Alias CBX; Cbx1; Cbx-rs2; chromobox 1; chromobox homolog 1; chromobox homolog 1 (HP1 beta homolog Drosophila ); chromobox protein homolog 1; E430007M08Rik; heterochromatin protein 1 homolog beta; heterochromatin protein 1-beta; heterochromatin protein p25; heterochromatin protein p25 beta; HGNC:1551; HP1 beta; HP1 beta homolog; HP1B; Hp1beta; HP1-BETA; HP1Hsbeta; HP1Hs-beta; M31; MOD1; modifier 1 protein; p25beta
Gene Symbols CBX1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 166-185. Recombinant protein control fragment (Product #RP-110054).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.