Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HP1BP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100562 Supplier Invitrogen™ Supplier No. PA5100562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HP1BP3.

HP1BP3 is the component of heterochromatin, may be involved in chromatin structure and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1BP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HP1BP3
Gene Accession No. Q3TEA8, Q5SSJ5, Q6P747
Gene Alias heterochromatin protein 1 binding protein 3; heterochromatin protein 1, binding protein 3; heterochromatin protein 1-binding protein 3; Hp1bp3; HP1BP74; HP1-BP74; Protein HP1-BP74
Gene Symbols HP1BP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human HP1BP3(Accession Q5SSJ5), corresponding to amino acid residues R307-K357.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15441, 313647, 50809
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.