Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HP1BP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565854 Supplier Invitrogen™ Supplier No. PA565854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITGKGASGTFQ LKKSGEKPLL GGSLMEYAIL SAIAAMNEPK TCSTTALKKY VLENHPGTNS NYQMHLLKKT LQKCEKNGWM E Highest antigen sequence identity to the following orthologs - mouse 98%, rat 91%.

HP1BP3 is the component of heterochromatin, may be involved in chromatin structure and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1BP3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HP1BP3
Gene Accession No. Q5SSJ5
Gene Alias heterochromatin protein 1 binding protein 3; heterochromatin protein 1, binding protein 3; heterochromatin protein 1-binding protein 3; Hp1bp3; HP1BP74; HP1-BP74; Protein HP1-BP74
Gene Symbols HP1BP3
Host Species Rabbit
Immunogen Recombinant Human HP1BP3. Recombinant protein control fragment (Product #RP-106522).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.