Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ HP35 Disposable Microtome Blades

Catalog No. p-4529783
Click to view available options
Coating:
Coated
This item is not returnable. View return policy
This item is not returnable. View return policy

Epredia™ HP35 microtome blades are coated for reliable and thin sectioning.

  • High-profile
  • Increased durability

Specifications

Product Type High Profile Blade
Profile High
For Use With (Application) Designed for high-quality thin sectioning
Disposable Single-use
Coating Coated
Cutting Angle 35°
Thickness (English) 0.01 in.
Thickness (Metric) 0.31 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.