Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPa1 Antibody (DHIC2-2C12), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$181.90 - $487.50
Specifications
Antigen | HPa1 |
---|---|
Clone | DHIC2-2C12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP118949SS
![]() |
Novus Biologicals
NBP118949SS |
0.025 mL |
Each for $181.90
|
|
|||||
NBP118949
![]() |
Novus Biologicals
NBP118949 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
HPa1 Monoclonal specifically detects HPa1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
HPa1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Pancreatic Stem Cell Markers, Stem Cell Markers | |
Tissue culture supernatant with 0.05% Sodium Azide | |
Human pancreatic enriched alpha cells | |
Primary | |
Store at 4C. Do not freeze. |
DHIC2-2C12 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
human pancreatic alpha cell | |
IgM | |
Tissue culture supernatant | |
Specific for alpha endocrine cell types. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title