Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Luciferase Antibody (Luci 21 1-107) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 18 publications
$236.98 - $591.43
Specifications
| Antigen | Luciferase |
|---|---|
| Clone | Luci 21 1-107 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:200. Use reported by customer review, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 31069316) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600307SS
![]() |
Novus Biologicals
NB600307SS |
0.025 mL |
Each for $236.98
|
|
|||||
NB600307
![]() |
Novus Biologicals
NB600307 |
0.1 mL |
Each for $591.43
|
|
|||||
Description
Luciferase Monoclonal specifically detects Luciferase in Firefly samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Luciferase | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins | |
| PBS with 0.02% Sodium Azide | |
| ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
| IgG1 κ | |
| Protein A or G purified | |
| This Luciferase Antibody (Luci 21 1-107) is specific for Luciferase, recognizing a peptide consisting of the first 258 amino acids. Further epitope mapping has not been done at this time. |
| Luci 21 1-107 | |
| Western Blot 1:1000, Flow Cytometry 1:200. Use reported by customer review, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 31069316) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Firefly | |
| P08659 | |
| This Luciferase Antibody (Luci 21 1-107) was developed against luciferase protein from Photinus pyralis (North American firefly). [UniProt# P08659]. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 61 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title