Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPa1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595775 Supplier Invitrogen™ Supplier No. PA595775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Jurkat, BxPC-3, A-431, mouse spleen, mouse lung, rat spleen, rat lung; Cellular Location: Lysosome membrane, Nucleus, Peripheral membrane protein Immunogen sequence: QDVVDLDFFT QEPLHLVSPS FLSVTIDANL ATDPRFLILL GSPKLRTLAR GLSPAYLRFG GTKTDFLIFD PKKESTFEER SYWQSQVNQD ICKYGSIPPD VEEKLRLEWP YQEQLLLREH YQKKFKNSTY SRSSVDVLYT FANCSGLDLI FGLNALLRTA DLQWNSSNAQ LLLDYCSSKG YNISWELGNE PNSFLKKADI FINGSQLGED FIQLHKLLRK STFKNAKLYG PDVGQPRRKT.

Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al. ). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPa1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene HPSE
Gene Accession No. Q6YGZ1, Q71RP1, Q9Y251
Gene Alias Endo-glucoronidase; endoglycosidase heparanase; HEP; Heparanase; Heparanase 50 kDa subunit; Heparanase 8 kDa subunit; Heparanase-1; heparanase-like; Hpa; HPA1; HPR1; HPSE; HPSE1; HSE1
Gene Symbols HPSE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 36-275 of human Heparanase 1 (NP_0010920101).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10855, 15442, 64537
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.