Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPa2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560773 Supplier Invitrogen™ Supplier No. PA560773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKRTDFLQFQ NLRNPAKSRG GPGPDYYLKN YEDDIVRSDV ALDKQKGCKI AQHPDVMLEL QREKAAQMHL VLLKQQFSNT Y Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 25%.

This gene encodes a heparanase enzyme. The encoded protein is a endoglycosidase that degrades heparin sulfate proteoglycans located on the extracellular matrix and cell surface. This protein may be involved in biological processes involving remodeling of the extracellular matrix including angiogenesis and tumor progression. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPa2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HPSE2
Gene Accession No. Q8WWQ2
Gene Alias Gm968; heparanase 2; heparanase 2 (inactive); heparanase 3; heparanase-2; heparanase-like protein; Hpa2; HPR2; Hpse2; Inactive heparanase-2; UFS; UFS1
Gene Symbols HPSE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HPa2. Recombinant protein control fragment (Product #RP-97170).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60495
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.