Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPa2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121181 Supplier Invitrogen™ Supplier No. PA5121181
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: BT-474, 22Rv1, HeLa, SKOV3, Mouse brain, Mouse small intestine, Mouse testis, Mouse uterus, Rat brain. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: GDRRPLPVDR AAGLKEKTLI LLDVSTKNPV RTVNENFLSL QLDPSIIHDG WLDFLSSKRL VTLARGLSPA FLRFGGKRTD FLQFQNLRNP AKSRGGPGPD YYLKNYEDDI VRSDVALDKQ KGCKIAQHPD VMLELQREKA AQMHLVLLKE QFSNTYSNLI LTARSLDKLY NFADCSGLHL IFALNALRRN PNNSWNSSSA L.

This gene encodes a heparanase enzyme. The encoded protein is a endoglycosidase that degrades heparin sulfate proteoglycans located on the extracellular matrix and cell surface. This protein may be involved in biological processes involving remodeling of the extracellular matrix including angiogenesis and tumor progression. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPa2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HPSE2
Gene Accession No. B2RY83, Q8WWQ2
Gene Alias Gm968; heparanase 2; heparanase 2 (inactive); heparanase 3; heparanase-2; heparanase-like protein; Hpa2; HPR2; Hpse2; Inactive heparanase-2; UFS; UFS1
Gene Symbols HPSE2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 42-242 of human HPSE2 (NP_068600.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 368128, 545291, 60495
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.