Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPCAL1 Mouse anti-Human, Unconjugated, Clone: 1E10, MilliporeSigma™

Catalog No. MABC1139100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1139-100 100 μg
MABC1139-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1139-100 Supplier MilliporeSigma Supplier No. MABC1139100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detects Hippocalcin-like protein 1 using this mouse monoclonal Anti-HPCAL1, clone 1E10, Cat. No. MABC1139, tested for use in Western Blotting and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPCAL1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 1E10
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in PBS without azide.
Gene Alias Hippocalcin-like protein 1;Calcium-binding protein BDR-1;HLP2;Visinin-like protein 3;VILIP-3
Gene Symbols HPCAL1;BDR1
Host Species Mouse
Immunogen Purified native and denatured human HPCAL1.
Molecular Weight of Antigen ∽22 kDa observed; 22.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_602293.1
Test Specificity Clone 1E10 specifically detects Hippocalcin-like protein 1 (HPCAL1) in human cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.