Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPCAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527536 Supplier Invitrogen™ Supplier No. PA527536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HPCAL1 tansfected 293T cell, mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (90%), Xenopus laevis (96%), Pig (99%), Chicken (98%), Sheep (99%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPCAL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene HPCAL1
Gene Accession No. P37235, P62748
Gene Alias BDR1; Calcium-binding protein BDR-1; hippocalcin like 1; hippocalcin-like 1; hippocalcin-like 1; visinin like 3; neural visinin-like 3; neural visinin-like protein 3; hippocalcin-like protein 1; HLP2; Hpcal1; neural visinin-like 3; neural visinin-like Ca2+-binding protein type 3; neural visinin-like protein 3; NVL-3; Nvp3; NVP-3; VILIP3; VILIP-3; visinin like 3; visinin-like protein 3; Vnsl3; Vsnl3
Gene Symbols HPCAL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HPCAL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3241, 53602
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.