Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPD Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299592B
Description
HPD Polyclonal specifically detects HPD in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HPD | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
4HPPDEC 1.13.11.27,4-HPPD, 4-hydroxyphenylpyruvate dioxygenase, GLOD3, glyoxalase domain containing 3,4-hydroxyphenylpyruvic acid oxidase, HPPDase, PPD | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human HPD. | |
0.1 mL | |
metabolism | |
3242 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction