Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPd1 Antibody (DHIC2-4A10), Novus Biologicals™
SDP

Catalog No. NBP118953SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118953SS 0.025 mL
NBP118953 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP118953SS Supplier Novus Biologicals Supplier No. NBP118953SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

HPd1 Monoclonal specifically detects HPd1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen HPd1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone DHIC2-4A10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100
Formulation Tissue culture supernatant with 0.09% Sodium Azide
Gene Alias DHIC2-4A10, HPd1, human pancreatic duct cell
Host Species Mouse
Immunogen Human pancreatic enriched duct cells
Purification Method Tissue culture supernatant
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cell Biology, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Specific for duct cell types.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.