Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPd2 Antibody (DHIC2-5H10), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$160.07 - $480.74
Specifications
| Antigen | HPd2 |
|---|---|
| Clone | DHIC2-5H10 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP118955SS
![]() |
Novus Biologicals
NBP118955SS |
0.025 mL |
Each for $160.07
|
|
|||||
NBP118955
![]() |
Novus Biologicals
NBP118955 |
0.1 mL |
Each for $480.74
|
|
|||||
Description
HPd2 Monoclonal specifically detects HPd2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| HPd2 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Pancreatic Stem Cell Markers, Stem Cell Markers | |
| Tissue culture supernatant with 0.05% Sodium Azide | |
| Human pancreatic enriched duct cells | |
| Primary | |
| Store at 4C. Do not freeze. |
| DHIC2-5H10 | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human | |
| human pancreatic duct cell | |
| IgM | |
| Tissue culture supernatant | |
| Specific for duct cell types. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title