Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPd2 Mouse anti-Human, Unconjugated, Clone: DHIC3-5H10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS2044100UG
Description
Anti-HPd2, clone DHIC3-5H10, Cat. No. MABS2044, is a mouse monoclonal antibody that detects Duct2 cells of pancreas and has been tested for use in Immunofluorescence and Immunohistochemistry (Paraffin).
Specifications
HPd2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide. | |
Mouse | |
Purified by ion-exchange chromatography | |
RUO | |
Primary | |
Clone DCHI3-5H10 specifically detects human pancreatic duct cells. | |
Stable for 1 year at 2-8°C from date of receipt. | |
Purified |
Immunohistochemistry (Paraffin), Immunofluorescence | |
DHIC3-5H10 | |
Unconjugated | |
Human pancreatics duct cells | |
Trypsin-dissociated human pancreatic duct cells. | |
100 μg | |
Signaling | |
NA | |
Human | |
Antibodies | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction