Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPd3 Mouse anti-Human, Unconjugated, Clone: DHIC5-4D9, MilliporeSigma™

Catalog No. MABS2040100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS2040-100 100 μg
MABS2040-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS2040-100 Supplier MilliporeSigma Supplier No. MABS2040100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HPd3, clone DHIC5-4D9, Cat. No. MABS2040, is a mouse monoclonal antibody that detects pancreatic and hepatic ductal cells and has been tested for use in Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPd3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DHIC5-4D9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide.
Gene Alias Human pancreas and liver duct cells
Host Species Mouse
Immunogen Human pancreas enriched in duct cells.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NA
Test Specificity Clone DHIC5-4D9 detects human pancreatic and liver duct cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.