Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPDL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556898
Description
Immunogen sequence: SPGEDPELGL EMTAGFGLGG LRLTALQAQP GSIVPTLVLA ESLPGATTRQ DQVEQFLARH KGPGLQHVGL YTPNIVEATE GVATAGGQFL AP Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.
May have dioxygenase activity.
Specifications
HPDL | |
Polyclonal | |
Unconjugated | |
HPDL | |
4-HPPD-L; 4-hydroxyphenylpyruvate dioxygenase like; 4-hydroxyphenylpyruvate dioxygenase-like protein; GLOXD1; glyoxalase domain containing 1; glyoxalase domain-containing protein 1; HPDL; RP4-534D1.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84842 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96IR7 | |
HPDL | |
Recombinant protein corresponding to Human HPDL. Recombinant protein control fragment (Product #RP-94520). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction