Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPGD Mouse anti-Human, Clone: OTI1C10, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA600099

Description
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.Specifications
HPGD | |
Monoclonal | |
0.5 mg/mL | |
PBS with <5% trehalose and 0.05% sodium azide | |
P15428 | |
HPGD | |
Full length human recombit protein of human HPGD produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
OTI1C10 | |
Unconjugated | |
HPGD | |
15-PGDH, PGDH, PGDH1, PHOAR1, SDR36C1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3248 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction