Learn More
Invitrogen™ HPGDS Monoclonal Antibody (735301)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | HPGDS |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 735301 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Hpgds |
| Gene Accession No. | O60760 |
| Gene Alias | EC 2.5.1.18; EC 5.3.99.2; glutathione S-transferase; glutathione S-transferase sigma; glutathione-dependent PGD synthase; glutathione-dependent PGD synthetase; glutathione-requiring prostaglandin D synthase; GST class-sigma; GSTS; GSTS1; GSTS1-1; hematopoietic prostaglandin D synthase; hematopoietic prostaglandin D2 synthase; Hpgds; H-PGDS; PGD2; PGDS; prostaglandin D2 synthase 2, hematopoietic; prostaglandin D2 synthase, hematopoietic; prostaglandin-H2 D-isomerase; Ptgds2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.