Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPGDS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554958
Description
Immunogen sequence: KQDVKEQMFN ELLTYNAPHL MQDLDTYLGG REWLIGNSVT WADFYWEICS TTLLVFKPDL LDNHPRLVTL RKKVQAIPAV ANWI Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
PGDS is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes. Prostaglandin-D synthase is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes.
Specifications
HPGDS | |
Polyclonal | |
Unconjugated | |
Hpgds | |
EC 2.5.1.18; EC 5.3.99.2; glutathione S-transferase; glutathione S-transferase sigma; glutathione-dependent PGD synthase; glutathione-dependent PGD synthetase; glutathione-requiring prostaglandin D synthase; GST class-sigma; GSTS; GSTS1; GSTS1-1; hematopoietic prostaglandin D synthase; hematopoietic prostaglandin D2 synthase; Hpgds; H-PGDS; PGD2; PGDS; prostaglandin D2 synthase 2, hematopoietic; prostaglandin D2 synthase, hematopoietic; prostaglandin-H2 D-isomerase; Ptgds2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27306 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60760 | |
Hpgds | |
Recombinant protein corresponding to Human HPGDS. Recombinant protein control fragment (Product #RP-90630). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction